Product Details

SNP ID
rs141746604
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:53036073 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCCTGGAACCCAGGACGGACCTG[G/T]AGCCCGGTGACCATCGGGATCGCGC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
POM121L12 PubMed Links

Gene Details

Gene
POM121L12
Gene Name
POM121 transmembrane nucleoporin like 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182595.3 418 Missense Mutation TGG,TGT W134C NP_872401.3

View Full Product Details