Product Details

SNP ID
rs141807671
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100576243 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTCCCGCCTCTGCTCACCTGGC[A/G]CAGCTGAGTCATTAAGTCCTTCTCA
Phenotype
MIM: 604019 MIM: 609097
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AGFG2 PubMed Links

Gene Details

Gene
AGFG2
Gene Name
ArfGAP with FG repeats 2
There are no transcripts associated with this gene.

Gene
FBXO24
Gene Name
F-box protein 24
There are no transcripts associated with this gene.

Gene
LRCH4
Gene Name
leucine rich repeats and calponin homology domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289934.1 1721 Missense Mutation CGC,TGC R545C NP_001276863.1
NM_002319.4 1721 Missense Mutation CGC,TGC R545C NP_002310.2
Gene
SAP25
Gene Name
Sin3A associated protein 25
There are no transcripts associated with this gene.

Gene
ZASP
Gene Name
ZO-2 associated speckle protein
There are no transcripts associated with this gene.

View Full Product Details