Product Details

SNP ID
rs142061761
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100430574 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACTCTGTGTTACCCTCCAACTTC[C/T]TCCTGGGGGGCAATATCTTTGATCC
Phenotype
MIM: 611478
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MEPCE PubMed Links

Gene Details

Gene
MEPCE
Gene Name
methylphosphate capping enzyme
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001194990.1 944 Intron NP_001181919.1
NM_001194991.1 944 Intron NP_001181920.1
NM_001194992.1 944 Intron NP_001181921.1
NM_019606.5 944 Missense Mutation CTC,TTC L186F NP_062552.2
XM_011516410.2 944 Intron XP_011514712.1
Gene
PPP1R35
Gene Name
protein phosphatase 1 regulatory subunit 35
There are no transcripts associated with this gene.

Gene
ZCWPW1
Gene Name
zinc finger CW-type and PWWP domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258008.1 944 Intron NP_001244937.1
NM_017984.4 944 Intron NP_060454.3

View Full Product Details