Product Details

SNP ID
rs142221015
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:39951813 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCCTAGTCCCTACAGCAGCAGCA[G/T]CTGGCGCCGCTCTCGCAGTCCCTAC
Phenotype
MIM: 603309
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CDK13 PubMed Links

Gene Details

Gene
CDK13
Gene Name
cyclin dependent kinase 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003718.4 2313 Missense Mutation AGC,ATC S391I NP_003709.3
NM_031267.3 2313 Missense Mutation AGC,ATC S391I NP_112557.2
XM_011515597.2 2313 Missense Mutation AGC,ATC S391I XP_011513899.1
XM_017012750.1 2313 Missense Mutation AGC,ATC S391I XP_016868239.1
XM_017012751.1 2313 Missense Mutation AGC,ATC S391I XP_016868240.1

View Full Product Details