Product Details

SNP ID
rs142231743
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:144095654 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACTTTTTCTGTCAAATCATGTCC[A/G]TATTCAAATTGGCCTGTGCTGACAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR2A12 PubMed Links

Gene Details

Gene
OR2A12
Gene Name
olfactory receptor family 2 subfamily A member 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004135.1 547 Missense Mutation ATA,GTA I183V NP_001004135.1

View Full Product Details