Product Details

SNP ID
rs143050451
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:103075881 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGTGGTCCAAGACCTCGCAGCCT[C/G]GTGTGTGTTTTGGAAATGGGAACAG
Phenotype
MIM: 611864 MIM: 609080
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ARMC10 PubMed Links

Gene Details

Gene
ARMC10
Gene Name
armadillo repeat containing 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161009.2 636 Intron NP_001154481.1
NM_001161010.2 636 Missense Mutation CAA,GAA Q82E NP_001154482.1
NM_001161011.2 636 Intron NP_001154483.1
NM_001161012.2 636 Intron NP_001154484.1
NM_001161013.2 636 Intron NP_001154485.1
NM_031905.4 636 Missense Mutation CAA,GAA Q82E NP_114111.2
XM_005250628.2 636 Missense Mutation CAA,GAA Q82E XP_005250685.1
XM_011516601.2 636 Intron XP_011514903.1
XM_011516602.2 636 Intron XP_011514904.1
XM_017012684.1 636 Missense Mutation CAT,GAT H82D XP_016868173.1
XM_017012685.1 636 Missense Mutation CAT,GAT H82D XP_016868174.1
XM_017012686.1 636 Missense Mutation CAT,GAT H82D XP_016868175.1
XM_017012687.1 636 Intron XP_016868176.1
XM_017012688.1 636 Missense Mutation CAT,GAT H82D XP_016868177.1
XM_017012689.1 636 Missense Mutation CAT,GAT H82D XP_016868178.1
Gene
FBXL13
Gene Name
F-box and leucine rich repeat protein 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001111038.1 636 Intron NP_001104508.1
NM_001287150.1 636 Intron NP_001274079.1
NM_145032.3 636 Intron NP_659469.3

View Full Product Details