Product Details

SNP ID
rs143067864
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:128457305 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGAACCTCTACCTGTTAGGTGTG[A/G]TACTGACCCTACTCTCCATCTTCGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HILPDA PubMed Links

Gene Details

Gene
HILPDA
Gene Name
hypoxia inducible lipid droplet associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098786.1 185 Missense Mutation ATA,GTA I13V NP_001092256.1
NM_013332.3 185 Missense Mutation ATA,GTA I13V NP_037464.1

View Full Product Details