Product Details
- SNP ID
-
rs143084654
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:127611096 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGCCATACAGTAGTGGGCAGCCAG[A/G]CCAGAGCAGGGCCTGAGAGCCACTA
- Phenotype
-
MIM: 615800
MIM: 167413
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FSCN3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs712701] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FSCN3
- Gene Name
- fascin actin-bundling protein 3
There are no transcripts associated with this gene.
- Gene
- PAX4
- Gene Name
- paired box 4
View Full Product Details