Product Details

SNP ID
rs143197576
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:47800738 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTCAGACTTGGAAAGGCCAAGGA[A/G]GCAGTCTTTTTGGCTTCTTCTGGGA
Phenotype
MIM: 609721
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C7orf69 PubMed Links

Gene Details

Gene
C7orf69
Gene Name
chromosome 7 open reading frame 69
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302627.1 8138 Intron NP_001289556.1
NM_025031.2 8138 Intron NP_079307.2
Gene
PKD1L1
Gene Name
polycystin 1 like 1, transient receptor potential channel interacting
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138295.3 8138 Missense Mutation CTC,TTC L2702F NP_612152.1
XM_017011798.1 8138 Missense Mutation CTC,TTC L2761F XP_016867287.1

View Full Product Details