Product Details

SNP ID
rs143226329
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:817599 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAAGCTGCCCATAATTGTGTCTT[A/C]TAAGCTTCAGTCATAGTATTGCCCA
Phenotype
MIM: 607723
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SUN1 PubMed Links

Gene Details

Gene
SUN1
Gene Name
Sad1 and UNC84 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130965.2 Intron NP_001124437.1
NM_001171944.1 Intron NP_001165415.1
NM_001171945.1 Intron NP_001165416.1
NM_001171946.1 Intron NP_001165417.1
NM_025154.5 Intron NP_079430.3

View Full Product Details