Product Details

SNP ID
rs143253409
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100057133 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCAAGTACCTTCCTAGCCTGGAG[A/G]TGTTCCGCCAGCGCTTCAGGCAGTT
Phenotype
MIM: 194510 MIM: 601261
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF3 PubMed Links

Gene Details

Gene
ZNF3
Gene Name
zinc finger protein 3
There are no transcripts associated with this gene.

Gene
ZSCAN21
Gene Name
zinc finger and SCAN domain containing 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145914.2 271 Missense Mutation ATG,GTG M43V NP_666019.1
XM_005250568.4 271 Missense Mutation ATG,GTG M43V XP_005250625.1
XM_006716113.3 271 Missense Mutation ATG,GTG M43V XP_006716176.1
XM_017012584.1 271 Missense Mutation ATG,GTG M43V XP_016868073.1
XM_017012585.1 271 Missense Mutation ATG,GTG M43V XP_016868074.1

View Full Product Details