Product Details

SNP ID
rs143352740
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:151467161 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGACTTGCCTTGTGAAGCTGCCCC[A/G]TCCATTTTTTCTGCCTCCAAAATTA
Phenotype
MIM: 601293
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RHEB PubMed Links

Gene Details

Gene
RHEB
Gene Name
Ras homolog enriched in brain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005614.3 980 Silent Mutation GAC,GAT D171D NP_005605.1
XM_011516457.1 980 Silent Mutation GAC,GAT D160D XP_011514759.1

View Full Product Details