Product Details

SNP ID
hCV173071952
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:130671680 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTACCATCTTTCCTCATATCTCT[C/G]TCATGGACTGGAGCAAAGGTGAGCT
Phenotype
MIM: 604355
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
COPG2 PubMed Links

Gene Details

Gene
COPG2
Gene Name
coatomer protein complex subunit gamma 2
There are no transcripts associated with this gene.

Gene
TSGA13
Gene Name
testis specific 13
There are no transcripts associated with this gene.

View Full Product Details