Product Details

SNP ID
rs143453856
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:132123417 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTGTTCTGAACTGCAAAATAGGA[A/T]TGCCTTATATTTACATACACAGGTA
Phenotype
MIM: 604280
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PLXNA4 PubMed Links

Gene Details

Gene
PLXNA4
Gene Name
plexin A4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105543.1 13467 Intron NP_001099013.1
NM_020911.1 13467 UTR 3 NP_065962.1
NM_181775.3 13467 Intron NP_861440.2
XM_005250686.4 13467 UTR 3 XP_005250743.1
XM_006716171.3 13467 UTR 3 XP_006716234.1
XM_011516676.2 13467 Intron XP_011514978.1
XM_017012779.1 13467 UTR 3 XP_016868268.1

View Full Product Details