Product Details

SNP ID
rs143560034
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:143292652 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTACGACTTGAGTCTCCCTTTTCC[A/G]GTGTGTGAGTCCTGTCCCCTTTACA
Phenotype
MIM: 600639 MIM: 616524
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CASP2 PubMed Links

Gene Details

Gene
CASP2
Gene Name
caspase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001224.4 424 Silent Mutation CCA,CCG P112P NP_001215.1
NM_032982.3 424 Silent Mutation CCA,CCG P143P NP_116764.2
NM_032983.3 424 Intron NP_116765.2
Gene
LOC105375545
Gene Name
uncharacterized LOC105375545
There are no transcripts associated with this gene.

Gene
TMEM139
Gene Name
transmembrane protein 139
There are no transcripts associated with this gene.

View Full Product Details