Product Details

SNP ID
rs143684679
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:134127420 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGACCAGAGCTGCCTCTCTCCTGA[C/G]AGGCTTGGGCAGATCCCGAACTGGA
Phenotype
MIM: 616478
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LRGUK PubMed Links

Gene Details

Gene
LRGUK
Gene Name
leucine rich repeats and guanylate kinase domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017011749.1 63 UTR 5 XP_016867238.1

View Full Product Details