Product Details

SNP ID
rs143788158
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:102467372 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACTGGGAGAAGTTCATGGCCACA[A/C]TGGGTCCTGAAGAGGAGGCTGAGAA
Phenotype
MIM: 613302 MIM: 615167 MIM: 604150
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ALKBH4 PubMed Links

Gene Details

Gene
ALKBH4
Gene Name
alkB homolog 4, lysine demethylase
There are no transcripts associated with this gene.

Gene
LRWD1
Gene Name
leucine rich repeats and WD repeat domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317721.1 682 Missense Mutation ATG,CTG M4L NP_001304650.1
NM_152892.2 682 Missense Mutation ATG,CTG M156L NP_690852.1
XM_017011845.1 682 UTR 5 XP_016867334.1
Gene
MIR4467
Gene Name
microRNA 4467
There are no transcripts associated with this gene.

Gene
MIR5090
Gene Name
microRNA 5090
There are no transcripts associated with this gene.

Gene
POLR2J
Gene Name
RNA polymerase II subunit J
There are no transcripts associated with this gene.

View Full Product Details