Product Details

SNP ID
rs143917001
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100102958 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTATACGAACTCCTGGATGAAGTG[C/T]TGGTGAGAATCAACAATCCCCTTCT
Phenotype
MIM: 602296 MIM: 600592 MIM: 612983 MIM: 612150 MIM: 612984 MIM: 602955
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AP4M1 PubMed Links

Gene Details

Gene
AP4M1
Gene Name
adaptor related protein complex 4 mu 1 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004722.3 535 Silent Mutation CTG,TTG L117L NP_004713.2
XM_005250689.4 535 Silent Mutation CTG,TTG L124L XP_005250746.1
XM_005250690.4 535 Intron XP_005250747.1
XM_006716175.3 535 Silent Mutation CTG,TTG L124L XP_006716238.1
XM_011516685.2 535 Silent Mutation CTG,TTG L124L XP_011514987.1
XM_017012790.1 535 UTR 5 XP_016868279.1
XM_017012791.1 535 Intron XP_016868280.1
Gene
MCM7
Gene Name
minichromosome maintenance complex component 7
There are no transcripts associated with this gene.

Gene
MIR106B
Gene Name
microRNA 106b
There are no transcripts associated with this gene.

Gene
MIR25
Gene Name
microRNA 25
There are no transcripts associated with this gene.

Gene
MIR93
Gene Name
microRNA 93
There are no transcripts associated with this gene.

Gene
TAF6
Gene Name
TATA-box binding protein associated factor 6
There are no transcripts associated with this gene.

View Full Product Details