Product Details

SNP ID
rs143942649
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:128835570 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGCGCCCTGGTGGACAACTGCGC[C/T]CCCGGTGAGTGGGCCAGTGAGCACA
Phenotype
MIM: 102565
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FLNC PubMed Links

Gene Details

Gene
FLNC
Gene Name
filamin C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127487.1 806 Silent Mutation GCC,GCT A199A NP_001120959.1
NM_001458.4 806 Silent Mutation GCC,GCT A199A NP_001449.3

View Full Product Details