Product Details

SNP ID
rs144089181
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:47994458 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACCCTGGGAACCTGGAAAAGCCC[A/G]GCACTTTCCAGGGTAGACATCCGGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SUN3 PubMed Links

Gene Details

Gene
SUN3
Gene Name
Sad1 and UNC84 domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001030019.1 805 Missense Mutation CGG,TGG R240W NP_001025190.1
NM_001284350.1 805 Missense Mutation CGG,TGG R228W NP_001271279.1
NM_152782.3 805 Missense Mutation CGG,TGG R240W NP_689995.3
XM_017011930.1 805 Missense Mutation CGG,TGG R240W XP_016867419.1

View Full Product Details