Product Details

SNP ID
rs144330000
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:44566064 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTGCTTGAAGCTGCGCAGTGGGT[G/T]CTGGGACTTTGCTCTCTAAGGAGGC
Phenotype
MIM: 608023
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DDX56 PubMed Links

Gene Details

Gene
DDX56
Gene Name
DEAD-box helicase 56
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257189.1 2105 Missense Mutation AAC,CAC N488H NP_001244118.1
NM_019082.3 2105 Missense Mutation AAC,CAC N528H NP_061955.1

View Full Product Details