Product Details

SNP ID
rs144488207
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:73705220 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATGAGTTCTTCCAGCTCCTCCTT[C/T]GTCTCTGGGGAGGTAGAAAGGGTGG
Phenotype
MIM: 186590 MIM: 615733
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR4284 PubMed Links

Gene Details

Gene
MIR4284
Gene Name
microRNA 4284
There are no transcripts associated with this gene.

Gene
STX1A
Gene Name
syntaxin 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001165903.1 284 Silent Mutation ACA,ACG T71T NP_001159375.1
NM_004603.3 284 Silent Mutation ACA,ACG T71T NP_004594.1
XM_017012567.1 284 Silent Mutation ACA,ACG T71T XP_016868056.1
Gene
WBSCR22
Gene Name
Williams-Beuren syndrome chromosome region 22
There are no transcripts associated with this gene.

View Full Product Details