Product Details

SNP ID
rs144835173
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:113878241 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCTTGTTGAATTACAAATATTTT[C/T]TGATTTCGTAGAAATAGGTTGGCTA
Phenotype
MIM: 600917
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PPP1R3A PubMed Links

Gene Details

Gene
PPP1R3A
Gene Name
protein phosphatase 1 regulatory subunit 3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002711.3 2428 Missense Mutation AAA,GAA K951E NP_002702.2
XM_005250473.3 2428 Missense Mutation AAA,GAA K750E XP_005250530.1

View Full Product Details