Product Details

SNP ID
rs145033896
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:80135166 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCTCTCGCTTTCGGCACCATGGG[A/C]TGCACGCTGAGCGCCGAGGACAAGG
Phenotype
MIM: 139310
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GNAI1 PubMed Links

Gene Details

Gene
GNAI1
Gene Name
G protein subunit alpha i1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256414.1 343 Intron NP_001243343.1
NM_002069.5 343 Silent Mutation GGA,GGC G2G NP_002060.4

View Full Product Details