Product Details

SNP ID
rs145085742
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:152648677 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAACCCCACTTTCTCCAATAATAA[A/C]AAAATGTTTTTTTAAACTGTTACTT
Phenotype
MIM: 600375
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
XRCC2 PubMed Links

Gene Details

Gene
XRCC2
Gene Name
X-ray repair cross complementing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005431.1 894 Missense Mutation GTT,TTT V270F NP_005422.1

View Full Product Details