Product Details

SNP ID
rs145211080
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:128457427 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTAGCCAACACAGAGCCCACCAA[A/G]GGCCTTCCAGACCATCCATCCAGAA
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
HILPDA PubMed Links

Gene Details

Gene
HILPDA
Gene Name
hypoxia inducible lipid droplet associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098786.1 307 Silent Mutation AAA,AAG K53K NP_001092256.1
NM_013332.3 307 Silent Mutation AAA,AAG K53K NP_037464.1

View Full Product Details