Product Details

SNP ID
rs145231926
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:140673623 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGCGGCGTCTTCCTGCATCTCC[A/G]CCTCTGGCTTCGCGCCGGCGCTCTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADCK2 PubMed Links

Gene Details

Gene
ADCK2
Gene Name
aarF domain containing kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052853.3 696 Missense Mutation CAC,CGC H98R NP_443085.2
XM_006716170.3 696 Missense Mutation CAC,CGC H98R XP_006716233.1
XM_011516675.2 696 Missense Mutation CAC,CGC H98R XP_011514977.1

View Full Product Details