Product Details

SNP ID
rs145337555
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:16861411 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAACATGATTCTAGGCACATATT[A/G]CCCATCAGGTGATAAATTCTTATCA
Phenotype
MIM: 609482
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AGR3 PubMed Links

Gene Details

Gene
AGR3
Gene Name
anterior gradient 3, protein disulphide isomerase family member
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_176813.4 392 Missense Mutation NP_789783.1
XM_005249627.2 392 Nonsense Mutation XP_005249684.1
XM_011515152.2 392 Nonsense Mutation XP_011513454.1

View Full Product Details