Product Details

SNP ID
rs145495721
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:98212657 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAGAAGCTCTCCAAGATCGAGAC[A/G]CTCACGCTGGCCAAGAACTACATCA
Phenotype
MIM: 608606 MIM: 610989 MIM: 614781
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BHLHA15 PubMed Links

Gene Details

Gene
BHLHA15
Gene Name
basic helix-loop-helix family member a15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_177455.3 404 Silent Mutation ACA,ACG T116T NP_803238.1
Gene
LMTK2
Gene Name
lemur tyrosine kinase 2
There are no transcripts associated with this gene.

Gene
TECPR1
Gene Name
tectonin beta-propeller repeat containing 1
There are no transcripts associated with this gene.

View Full Product Details