Product Details

SNP ID
rs145582437
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:97853183 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGTATCAGATTGGAATCCTCAAA[C/T]GTCTCTCTCAGGAGATGTTTTTCTA
Phenotype
MIM: 108370
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ASNS PubMed Links

Gene Details

Gene
ASNS
Gene Name
asparagine synthetase (glutamine-hydrolyzing)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178075.1 1229 Intron NP_001171546.1
NM_001178076.1 1229 Intron NP_001171547.1
NM_001178077.1 1229 Silent Mutation ACA,ACG T368T NP_001171548.1
NM_001673.4 1229 Intron NP_001664.3
NM_133436.3 1229 Silent Mutation ACA,ACG T451T NP_597680.2
NM_183356.3 1229 Intron NP_899199.2

View Full Product Details