Product Details

SNP ID
rs145588023
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:148590962 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTCAAAGCCTCAGCCTTGAAGAG[C/T]GTCCCTGGAGACTTCCAGGCCCCCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C7orf33 PubMed Links

Gene Details

Gene
C7orf33
Gene Name
chromosome 7 open reading frame 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145304.3 398 Missense Mutation CGT,TGT R13C NP_660347.1

View Full Product Details