Product Details

SNP ID
rs145693200
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:143288510 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCTTCCAGCACAAGGAGCTGATG[A/G]CCGCTGACAGGGGACGCAGGTAAGT
Phenotype
MIM: 600639 MIM: 616524
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CASP2 PubMed Links

Gene Details

Gene
CASP2
Gene Name
caspase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001224.4 296 Intron NP_001215.1
NM_032982.3 296 Missense Mutation ACC,GCC T19A NP_116764.2
NM_032983.3 296 Missense Mutation ACC,GCC T19A NP_116765.2
Gene
LOC105375545
Gene Name
uncharacterized LOC105375545
There are no transcripts associated with this gene.

Gene
TMEM139
Gene Name
transmembrane protein 139
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242773.2 296 Intron NP_001229702.1
NM_001242774.2 296 Intron NP_001229703.1
NM_001242775.2 296 Intron NP_001229704.1
NM_001282876.1 296 Intron NP_001269805.1
NM_001282877.1 296 Intron NP_001269806.1
NM_153345.3 296 Intron NP_699176.1

View Full Product Details