Product Details

SNP ID
rs145773939
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:141742216 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAGTCCGAAACAACTACCAGTTT[G/T]GTTCTTGAAAGATGTAAGTAGCTAA
Phenotype
MIM: 600439
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SSBP1 PubMed Links

Gene Details

Gene
SSBP1
Gene Name
single stranded DNA binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256510.1 333 Missense Mutation TTG,TTT L24F NP_001243439.1
NM_001256511.1 333 Missense Mutation TTG,TTT L24F NP_001243440.1
NM_001256512.1 333 Missense Mutation TTG,TTT L24F NP_001243441.1
NM_001256513.1 333 Intron NP_001243442.1
NM_003143.2 333 Intron NP_003134.1
XM_005250048.4 333 Missense Mutation TTG,TTT L24F XP_005250105.1
XM_005250049.4 333 Missense Mutation TTG,TTT L24F XP_005250106.1
XM_005250050.4 333 Missense Mutation TTG,TTT L24F XP_005250107.1
XM_005250051.4 333 Missense Mutation TTG,TTT L24F XP_005250108.1
Gene
WEE2-AS1
Gene Name
WEE2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details