Product Details
- SNP ID
-
rs145780869
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:103083830 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACAATGCAGCCTTTTCAGTTAACCA[A/G]GTAAGTACCTCAACTCAGCAAGCAA
- Phenotype
-
MIM: 611864
MIM: 609080
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ARMC10
PubMed Links
Gene Details
- Gene
- ARMC10
- Gene Name
- armadillo repeat containing 10
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001161009.2 |
785 |
Silent Mutation |
CAA,CAG |
Q96Q |
NP_001154481.1 |
NM_001161010.2 |
785 |
Silent Mutation |
CAA,CAG |
Q131Q |
NP_001154482.1 |
NM_001161011.2 |
785 |
Silent Mutation |
CAA,CAG |
Q96Q |
NP_001154483.1 |
NM_001161012.2 |
785 |
Silent Mutation |
CAA,CAG |
Q96Q |
NP_001154484.1 |
NM_001161013.2 |
785 |
Silent Mutation |
CAA,CAG |
Q96Q |
NP_001154485.1 |
NM_031905.4 |
785 |
Silent Mutation |
CAA,CAG |
Q131Q |
NP_114111.2 |
XM_005250628.2 |
785 |
Silent Mutation |
CAA,CAG |
Q131Q |
XP_005250685.1 |
XM_011516601.2 |
785 |
Silent Mutation |
CAA,CAG |
Q104Q |
XP_011514903.1 |
XM_011516602.2 |
785 |
Intron |
|
|
XP_011514904.1 |
XM_017012684.1 |
785 |
Silent Mutation |
CAA,CAG |
Q140Q |
XP_016868173.1 |
XM_017012685.1 |
785 |
Silent Mutation |
CAA,CAG |
Q140Q |
XP_016868174.1 |
XM_017012686.1 |
785 |
Silent Mutation |
CAA,CAG |
Q140Q |
XP_016868175.1 |
XM_017012687.1 |
785 |
Intron |
|
|
XP_016868176.1 |
XM_017012688.1 |
785 |
Silent Mutation |
CAA,CAG |
Q140Q |
XP_016868177.1 |
XM_017012689.1 |
785 |
Silent Mutation |
CAA,CAG |
Q140Q |
XP_016868178.1 |
- Gene
- FBXL13
- Gene Name
- F-box and leucine rich repeat protein 13
There are no transcripts associated with this gene.
View Full Product Details