Product Details

SNP ID
rs146165353
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:106093076 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTATTTTAATATCTGTCAGAGCT[G/T]TAGCCCAGGCTGAAGTGCTCACCAA
Phenotype
MIM: 616665
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SYPL1 PubMed Links

Gene Details

Gene
SYPL1
Gene Name
synaptophysin like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006754.3 636 Missense Mutation AAA,ACA K173T NP_006745.1
NM_182715.2 636 Missense Mutation AAA,ACA K155T NP_874384.1

View Full Product Details