Product Details

SNP ID
rs146256377
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:155071244 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGCTAGGTCGGAGGCTGTGCCA[A/G]CTTTGGATCGCGTGCGACGTGCTTT
Phenotype
MIM: 601305
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HTR5A PubMed Links

Gene Details

Gene
HTR5A
Gene Name
5-hydroxytryptamine receptor 5A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024012.3 921 Silent Mutation CAA,CAG Q115Q NP_076917.1
Gene
HTR5A-AS1
Gene Name
HTR5A antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details