Product Details

SNP ID
rs146389993
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100488190 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCCCAGAGGGCCGAGAGTCCAGC[C/T]GGAAGGTTCCTCCGCAGAAGCCCAG
Phenotype
MIM: 615477 MIM: 611914
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NYAP1 PubMed Links

Gene Details

Gene
NYAP1
Gene Name
neuronal tyrosine phosphorylated phosphoinositide-3-kinase adaptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173564.3 683 Silent Mutation CGG,TGG R157W NP_775835.2
XM_006715907.3 683 Silent Mutation CGG,TGG R157W XP_006715970.1
XM_017011868.1 683 Silent Mutation CGG,TGG R157W XP_016867357.1
Gene
TSC22D4
Gene Name
TSC22 domain family member 4
There are no transcripts associated with this gene.

View Full Product Details