Product Details
- SNP ID
-
rs146500782
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:140086575 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGAAGGCTACAGTATTAAGATTAC[A/G]TTACGTTCAACAGCAACAGAATGAG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KDM7A
PubMed Links
Gene Details
- Gene
- KDM7A
- Gene Name
- lysine demethylase 7A
View Full Product Details