Product Details

SNP ID
rs146575754
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:127595498 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGGACCCTCCAGTGCCTAATCTC[C/T]GGTCGTTATTTGGAGTCCAATGGCA
Phenotype
MIM: 103188 MIM: 615800 MIM: 607418
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARF5 PubMed Links

Gene Details

Gene
ARF5
Gene Name
ADP ribosylation factor 5
There are no transcripts associated with this gene.

Gene
FSCN3
Gene Name
fascin actin-bundling protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020369.2 555 Silent Mutation TCC,TCT S112S NP_065102.1
Gene
GCC1
Gene Name
GRIP and coiled-coil domain containing 1
There are no transcripts associated with this gene.

View Full Product Details