Product Details

SNP ID
rs146605020
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:151234041 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTGAGCTCCCTGTTGGCTCTGCT[A/G]CGGCCAGCGCTTCCCCTCATCTTAG
Phenotype
MIM: 612510 MIM: 608037 MIM: 615245 MIM: 601737
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABCF2 PubMed Links

Gene Details

Gene
ABCF2
Gene Name
ATP binding cassette subfamily F member 2
There are no transcripts associated with this gene.

Gene
CHPF2
Gene Name
chondroitin polymerizing factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284295.1 1553 Intron NP_001271224.1
NM_019015.2 1553 Silent Mutation CTA,CTG L10L NP_061888.1
Gene
MIR671
Gene Name
microRNA 671
There are no transcripts associated with this gene.

Gene
SMARCD3
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 3
There are no transcripts associated with this gene.

View Full Product Details