Product Details

SNP ID
rs146672512
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100170483 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTCCAGAGCCGCTGGCATCCTCA[C/T]CTGCAAGGAAGAAGAGGGACAGAGC
Phenotype
MIM: 608235 MIM: 608489
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GAL3ST4 PubMed Links

Gene Details

Gene
GAL3ST4
Gene Name
galactose-3-O-sulfotransferase 4
There are no transcripts associated with this gene.

Gene
GPC2
Gene Name
glypican 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152742.2 1714 Missense Mutation GAT,GGT D496G NP_689955.1
Gene
STAG3
Gene Name
stromal antigen 3
There are no transcripts associated with this gene.

View Full Product Details