Product Details

SNP ID
rs146676508
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:44062813 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAGCCTCCATGGCCTTCCGCACC[A/G]TTTCCTCATCACCCAGGAACTGCAT
Phenotype
MIM: 610106 MIM: 612931 MIM: 606344
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DBNL PubMed Links

Gene Details

Gene
DBNL
Gene Name
drebrin like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014436.2 771 Intron NP_001014436.1
NM_001122956.1 771 Intron NP_001116428.1
NM_001284313.1 771 Intron NP_001271242.1
NM_001284315.1 771 Intron NP_001271244.1
NM_014063.6 771 Intron NP_054782.2
Gene
PGAM2
Gene Name
phosphoglycerate mutase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000290.3 771 Missense Mutation ACG,ATG T238M NP_000281.2
Gene
POLM
Gene Name
DNA polymerase mu
There are no transcripts associated with this gene.

View Full Product Details