Product Details

SNP ID
rs146750525
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:99621456 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCAGGCATCCAGCTGGGGCCAGT[G/T]GAGGTCAAGCCTGAATGGGGGATGC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZSCAN25 PubMed Links

Gene Details

Gene
ZSCAN25
Gene Name
zinc finger and SCAN domain containing 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145115.2 1053 Silent Mutation GTG,GTT V157V NP_660090.2
XM_005250194.2 1053 Silent Mutation GTG,GTT V157V XP_005250251.1
XM_011515905.2 1053 Silent Mutation GTG,GTT V157V XP_011514207.1
XM_011515906.2 1053 Silent Mutation GTG,GTT V157V XP_011514208.1
XM_011515907.2 1053 Silent Mutation GTG,GTT V157V XP_011514209.1
XM_011515908.2 1053 UTR 5 XP_011514210.1
XM_011515909.2 1053 Silent Mutation GTG,GTT V157V XP_011514211.1
XM_011515910.2 1053 Silent Mutation GTG,GTT V157V XP_011514212.1
XM_017011824.1 1053 Silent Mutation GTG,GTT V157V XP_016867313.1
XM_017011825.1 1053 UTR 5 XP_016867314.1
XM_017011826.1 1053 UTR 5 XP_016867315.1
XM_017011827.1 1053 Silent Mutation GTG,GTT V157V XP_016867316.1

View Full Product Details