Product Details

SNP ID
rs146769713
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:16600360 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATATAAGTTATGATTTTTCTAAC[C/T]ACCCTCTGTAGCTATAAATTTTTCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKMY2 PubMed Links

Gene Details

Gene
ANKMY2
Gene Name
ankyrin repeat and MYND domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020319.2 2098 UTR 3 NP_064715.1
XM_011515458.1 2098 UTR 3 XP_011513760.1

View Full Product Details