Product Details
- SNP ID
-
rs146805332
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:140337327 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAAACCCACATCCATCCTAGCTTG[C/T]CCCGGATCAGAGACACTAAATACTG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SLC37A3
PubMed Links
Gene Details
- Gene
- SLC37A3
- Gene Name
- solute carrier family 37 member 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001287498.1 |
1850 |
Missense Mutation |
GAC,GGC |
D434G |
NP_001274427.1 |
NM_032295.3 |
1850 |
Silent Mutation |
GGA,GGG |
G349G |
NP_115671.1 |
NM_207113.2 |
1850 |
Missense Mutation |
GAC,GGC |
D450G |
NP_996996.1 |
XM_011516626.2 |
1850 |
Silent Mutation |
GGA,GGG |
G399G |
XP_011514928.1 |
XM_011516627.2 |
1850 |
Silent Mutation |
GGA,GGG |
G383G |
XP_011514929.1 |
XM_017012712.1 |
1850 |
Missense Mutation |
GAC,GGC |
D450G |
XP_016868201.1 |
XM_017012713.1 |
1850 |
Silent Mutation |
GGA,GGG |
G349G |
XP_016868202.1 |
XM_017012714.1 |
1850 |
Missense Mutation |
GAC,GGC |
D369G |
XP_016868203.1 |
XM_017012715.1 |
1850 |
Missense Mutation |
GAC,GGC |
D362G |
XP_016868204.1 |
XM_017012716.1 |
1850 |
Silent Mutation |
GGA,GGG |
G261G |
XP_016868205.1 |
XM_017012717.1 |
1850 |
Missense Mutation |
GAC,GGC |
D302G |
XP_016868206.1 |
XM_017012718.1 |
1850 |
Intron |
|
|
XP_016868207.1 |
View Full Product Details