Product Details

SNP ID
rs146918232
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:150687083 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATGGACCAAAATGAACACAGTCA[C/G]TGGGGTAAGAAGGTGACTTCACGTG
Phenotype
MIM: 608085
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GIMAP2 PubMed Links

Gene Details

Gene
GIMAP2
Gene Name
GTPase, IMAP family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015660.2 112 Missense Mutation CAC,CAG H8Q NP_056475.1

View Full Product Details