Product Details

SNP ID
rs146939344
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:143026226 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATCTTTCAGGAGTTGACAGCAGC[A/G]TTTCATCCCATCTCGGAGGGCCTCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR9A2 PubMed Links

Gene Details

Gene
OR9A2
Gene Name
olfactory receptor family 9 subfamily A member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001658.1 907 Missense Mutation CGC,TGC R303C NP_001001658.1

View Full Product Details