Product Details

SNP ID
rs147074853
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:73831555 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAAAGCGCCAAGGCCAAGACCAT[A/G]ATCGTGGCGGGCGTGGTGTTCCTGT
Phenotype
MIM: 602909 MIM: 612546
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLDN4 PubMed Links

Gene Details

Gene
CLDN4
Gene Name
claudin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001305.4 693 Missense Mutation ATA,ATG I118M NP_001296.1
Gene
WBSCR27
Gene Name
Williams Beuren syndrome chromosome region 27
There are no transcripts associated with this gene.

View Full Product Details