Product Details
- SNP ID
-
rs147192522
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:141572776 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGATTTTATATTCTGTTTTAGATA[A/C]GTTGAGTTTGAAACCCCGACAGGCT
- Phenotype
-
MIM: 610345
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
AGK
PubMed Links
Gene Details
- Gene
- AGK
- Gene Name
- acylglycerol kinase
View Full Product Details