Product Details

SNP ID
rs147305510
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:73596373 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGGATGGTGCCCACCTTGTTGCT[A/G]TCTGGACGGCCCCGGCTGAGGATGG
Phenotype
MIM: 605678
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MLXIPL PubMed Links

Gene Details

Gene
MLXIPL
Gene Name
MLX interacting protein like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032951.2 1977 Silent Mutation GAC,GAT D643D NP_116569.1
NM_032952.2 1977 Silent Mutation GAC,GAT D643D NP_116570.1
NM_032953.2 1977 Silent Mutation GAC,GAT D643D NP_116571.1
NM_032954.2 1977 Silent Mutation GAC,GAT D643D NP_116572.1
XM_011516277.1 1977 Silent Mutation GAC,GAT D708D XP_011514579.1
XM_011516278.1 1977 Silent Mutation GAC,GAT D707D XP_011514580.1
XM_011516279.1 1977 Silent Mutation GAC,GAT D708D XP_011514581.1
XM_011516281.2 1977 Silent Mutation GAC,GAT D367D XP_011514583.1
XM_017012263.1 1977 Silent Mutation GAC,GAT D340D XP_016867752.1

View Full Product Details